4-Fluoroisoindoline 95%

Lieferant: Apollo Scientific

PC905110-1G
APOSPC905110-1GEA 270 EUR
APOSPC905110-1G
4-Fluoroisoindoline 95%
4-Fluoroisoindoline
Formel: C₈H₈FN
Molecular Weight: 137,154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027772
CAS-Nummer: 127168-78-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR