2-Bromo-5-chloro-3-fluoroisonicotinic acid 97%

Lieferant: Apollo Scientific
Achtung

PC905149-250MG
APOSPC905149-250MGEA 98.6 EUR
APOSPC905149-250MG
2-Bromo-5-chloro-3-fluoroisonicotinic acid 97%
2-Bromo-5-chloro-3-fluoroisonicotinic acid
Formel: C₆H₂BrClFNO₂
Molecular Weight: 254,441 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD20921503
CAS-Nummer: 514798-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR