4-Fluoro-1H-indol-6-amine 97%

Lieferant: Apollo Scientific
Achtung

PC905163-250MG
APOSPC905163-250MGEA 128 EUR
APOSPC905163-250MG
4-Fluoro-1H-indol-6-amine 97%
4-Fluoro-1H-indol-6-amine
Formel: C₈H₇FN₂
Molecular Weight: 150,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07357295
CAS-Nummer: 885518-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR