2-Fluoro-5-hydroxy-3-methoxypyridine 95%

Lieferant: Apollo Scientific

PC905174-1G
APOSPC905174-1GEA 806 EUR
APOSPC905174-1G
2-Fluoro-5-hydroxy-3-methoxypyridine 95%
2-Fluoro-5-hydroxy-3-methoxypyridine
Formel: C₆H₆FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16611061
CAS-Nummer: 1227511-69-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR