6-Fluoro-3-hydroxy-2-methylpyridine 97%

Lieferant: Apollo Scientific

PC905297-1G
APOSPC905297-1GEA 261 EUR
APOSPC905297-1G
6-Fluoro-3-hydroxy-2-methylpyridine 97%
6-Fluoro-3-hydroxy-2-methylpyridine
Formel: C₆H₆FNO
Molecular Weight: 127,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16611088
CAS-Nummer: 1227577-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR