2-Chloro-5-(trifluoromethyl)nicotinonitrile 98%

Lieferant: Apollo Scientific
Achtung

PC905310-1G PC905310-250MG
APOSPC905310-1GEA 243 EUR
APOSPC905310-1G APOSPC905310-250MG
2-Chloro-5-(trifluoromethyl)nicotinonitrile 98%
2-Chloro-5-(trifluoromethyl)nicotinonitrile
Formel: C₇H₂ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10697685
CAS-Nummer: 624734-22-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR