4-Fluoro-3-nitrophenylacetonitrile 95%

Lieferant: Apollo Scientific

PC905355-1G
APOSPC905355-1GEA 91.5 EUR
APOSPC905355-1G
4-Fluoro-3-nitrophenylacetonitrile 95%
4-Fluoro-3-nitrophenylacetonitrile
Formel: C₈H₅FN₂O₂
Molecular Weight: 180,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09909410
CAS-Nummer: 157662-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR