5-Fluor-2-methyl-1-indanon 95%
Lieferant: Apollo Scientific
Synonyme:
5-Fluoro-2-methyl-2,3-dihydro-1H-inden-1-one
PC905377-1G
APOSPC905377-1GEA
499
EUR
APOSPC905377-1G
5-Fluor-2-methyl-1-indanon 95%
5-Fluor-2-methyl-1-indanon
Formel:
C₁₀H₉FO Lagertemperatur: Raumtemperatur |
MDL:
MFCD07787592 CAS-Nummer: 41201-58-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...