6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-amine 95%

Lieferant: Apollo Scientific

PC905429-250MG
APOSPC905429-250MGEA 447 EUR
APOSPC905429-250MG
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-amine 95%
6-(Trifluoromethyl)imidazo[1,2-a]pyridin-2-amine
Formel: C₈H₆F₃N₃
Lagertemperatur: Gefrierschrank
MDL: MFCD11846602
CAS-Nummer: 1005785-87-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR