6-Fluoroquinoline-2-carboxylic acid 96%

Lieferant: Apollo Scientific
Achtung

PC905441-250MG
APOSPC905441-250MGEA 108 EUR
APOSPC905441-250MG
6-Fluoroquinoline-2-carboxylic acid 96%
6-Fluoroquinoline-2-carboxylic acid
Formel: C₁₀H₆FNO₂
Molecular Weight: 191,159 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09837268
CAS-Nummer: 86324-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR