(Perfluoropropyl)phenyliodonium trifluoromethanesulphonate 97%

Lieferant: Apollo Scientific

Synonyme: (Heptafluoropropyl)phenyliodonium Trifluoromethanesulphonate

PC905444-1G
APOSPC905444-1GEA 357 EUR
APOSPC905444-1G
(Perfluoropropyl)phenyliodonium trifluoromethanesulphonate 97%
(Perfluoropropyl)phenyliodonium trifluoromethanesulphonate
Formel: C₁₀H₅F₁₀IO₃S
Molecular Weight: 522,10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135121
CAS-Nummer: 77758-79-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR