2-(3-(Trifluoromethyl)phenyl)ethanol 97%

Lieferant: Apollo Scientific

PC905462-5G
APOSPC905462-5GEA 143 EUR
APOSPC905462-5G
2-(3-(Trifluoromethyl)phenyl)ethanol 97%
2-(3-(Trifluoromethyl)phenyl)ethanol
Formel: C₉H₉F₃O
Molecular Weight: 190,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009710
CAS-Nummer: 455-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR