Potassium cyclopentoxymethyltrifluoroborate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Potassium ((cyclopentyloxy)methyl)trifluoroborate
PC905473-1G
PC905473-5G
APOSPC905473-1GEA
57
EUR
APOSPC905473-1G
APOSPC905473-5G
Potassium cyclopentoxymethyltrifluoroborate 95%
Potassium cyclopentoxymethyltrifluoroborate
Formel:
C₆H₁₁BF₃O·K Molecular Weight: 206.055 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD11505926 CAS-Nummer: 1027642-31-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...