2-(Chloromethyl)-6-(trifluoromethyl)pyridine 95%

Lieferant: Apollo Scientific

PC905474-1G
APOSPC905474-1GEA 522 EUR
APOSPC905474-1G
2-(Chloromethyl)-6-(trifluoromethyl)pyridine 95%
2-(Chloromethyl)-6-(trifluoromethyl)pyridine
Formel: C₇H₅ClF₃N
Molecular Weight: 195,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188826
CAS-Nummer: 849094-03-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR