2,6-Dimethyl-4-fluorbenzonitril 95%
Lieferant: Apollo Scientific
Synonyme:
4-Fluoro-2,6-dimethylbenzonitrile
PC905491-1G
APOSPC905491-1GEA
120
EUR
APOSPC905491-1G
2,6-Dimethyl-4-fluorbenzonitril 95%
2,6-Dimethyl-4-fluorbenzonitril
Formel:
C₉H₈FN Lagertemperatur: Raumtemperatur |
MDL:
MFCD04972839 CAS-Nummer: 14659-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...