6-Chloro-5-fluoro-2-oxindole 95%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-5-fluoroindolin-2-one

PC905563-5G
APOSPC905563-5GEA 115 EUR
APOSPC905563-5G
6-Chloro-5-fluoro-2-oxindole 95%
6-Chloro-5-fluoro-2-oxindole
Formel: C₈H₅ClFNO
Molecular Weight: 185.583 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09839325
CAS-Nummer: 100487-74-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR