3-Ethenylazetidine trifluoroacetic acid
Lieferant: Apollo Scientific
PC905643-1G
PC905643-5G
APOSPC905643-1GEA
655
EUR
APOSPC905643-1G
APOSPC905643-5G
3-Ethenylazetidine trifluoroacetic acid
3-Ethenylazetidine trifluoroacetic acid
Formel:
C₅H₉N·CF₃CO₂H Molecular Weight: 197.155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28166293 CAS-Nummer: 1630907-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...