3-Amino-4-fluorophenyl acetate 95%

Lieferant: Apollo Scientific

PC905928-5G
APOSPC905928-5GEA 1910 EUR
APOSPC905928-5G
3-Amino-4-fluorophenyl acetate 95%
3-Amino-4-fluorophenyl acetate
Formel: C₈H₈FNO₂
Molecular Weight: 169,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28099391
CAS-Nummer: 196610-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR