4-Bromo-2-Fluorobenzylamine 98%
Lieferant: Apollo Scientific
Synonyme:
(4-Bromo-2-fluorophenyl)methanamine
PC9062-25G
APOSPC9062-25GEA
140
EUR
APOSPC9062-25G
4-Bromo-2-Fluorobenzylamine 98%
4-Bromo-2-Fluorobenzylamine
Formel:
C₇H₇BrFN Molecular Weight: 204.042 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153076 CAS-Nummer: 112734-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...