Methyl 3-fluoroazetidine-3-carboxylate hydrochloride

Lieferant: Apollo Scientific

PC906219-1G
APOSPC906219-1GEA 923 EUR
APOSPC906219-1G
Methyl 3-fluoroazetidine-3-carboxylate hydrochloride
Methyl 3-fluoroazetidine-3-carboxylate hydrochloride
Formel: C₅H₈FNO₂·ClH
Molecular Weight: 169.582 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27988017
CAS-Nummer: 1421920-61-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR