tert-Butyl 3-fluoro-3-(fluoromethyl)azetidine-1-carboxylate

Lieferant: Apollo Scientific

PC906268-1G
APOSPC906268-1GEA 1370 EUR
APOSPC906268-1G
tert-Butyl 3-fluoro-3-(fluoromethyl)azetidine-1-carboxylate
tert-Butyl 3-fluoro-3-(fluoromethyl)azetidine-1-carboxylate
Formel: C₉H₁₅F₂NO₂
Molecular Weight: 207,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28133687
CAS-Nummer: 1466514-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR