2-Chloro-5-fluoro-3-methylbenzoic acid 97%

Lieferant: Apollo Scientific

PC906340-1G PC906340-5G
APOSPC906340-1GEA 219 EUR
APOSPC906340-1G APOSPC906340-5G
2-Chloro-5-fluoro-3-methylbenzoic acid 97%
2-Chloro-5-fluoro-3-methylbenzoic acid
Formel: C₈H₆ClFO
Molecular Weight: 188.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23712098
CAS-Nummer: 1427416-28-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR