N-(4-Fluorphenyl)formamid 95%

Lieferant: Apollo Scientific

Synonyme: 4'-Fluoroformanilide, 1-Fluoro-4-formamidobenzene

PC906379-1G PC906379-5G
APOSPC906379-1GEA 84.1 EUR
APOSPC906379-1G APOSPC906379-5G
N-(4-Fluorphenyl)formamid 95%
N-(4-Fluorphenyl)formamid
Formel: C₇H₆FNO
Molecular Weight: 139.13 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 459-25-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR