Methyl-3-fluor-2-thiophencarboxylat 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-fluorothiophene-2-carboxylate
PC906382-5G
APOSPC906382-5GEA
127
EUR
APOSPC906382-5G
Methyl-3-fluor-2-thiophencarboxylat 95%
Methyl-3-fluor-2-thiophencarboxylat
Formel:
C₆H₅FO₂S Molecular Weight: 160.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16606491 CAS-Nummer: 100421-52-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...