2-Chloro-4-fluoro-3-methylbenzoic acid 98%
Lieferant: Apollo Scientific
PC906389-250MG
APOSPC906389-250MGEA
68.3
EUR
APOSPC906389-250MG
2-Chloro-4-fluoro-3-methylbenzoic acid 98%
2-Chloro-4-fluoro-3-methylbenzoic acid
Formel:
C₈H₆ClFO₂ Molecular Weight: 188.59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846444 CAS-Nummer: 173315-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...