(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate 98%

Lieferant: Apollo Scientific

Synonyme: (Tridecafluorohexyl)phenyliodonium Trifluoromethanesulphonate

PC906432-1G
APOSPC906432-1GEA 306 EUR
APOSPC906432-1G
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate 98%
(Perfluorohexyl)phenyliodonium trifluoromethanesulphonate
Formel: C₁₃H₅F₁₆IO₃S
Molecular Weight: 672,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00135119
CAS-Nummer: 77758-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR