3-Fluoro-5-methoxypyridine 95%

Lieferant: Apollo Scientific

PC906458-1G
APOSPC906458-1GEA 560 EUR
APOSPC906458-1G
3-Fluoro-5-methoxypyridine 95%
3-Fluoro-5-methoxypyridine
Formel: C₆H₆FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD13185606
CAS-Nummer: 1060801-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR