4-Fluoroisoindoline-1,3-dione 90%

Lieferant: Apollo Scientific

PC906500-1G
APOSPC906500-1GEA 56.7 EUR
APOSPC906500-1G
4-Fluoroisoindoline-1,3-dione 90%
4-Fluoroisoindoline-1,3-dione
Formel: C₈H₄FNO₂
Molecular Weight: 165,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196660
CAS-Nummer: 51108-29-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR