2-Chloro-3,4-difluoroaniline 95%
Lieferant: Apollo Scientific
PC906534-25G
APOSPC906534-25GEA
1140
EUR
APOSPC906534-25G
2-Chloro-3,4-difluoroaniline 95%
2-Chloro-3,4-difluoroaniline
Formel:
C₆H₄ClF₂N Molecular Weight: 163,552 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044823 CAS-Nummer: 36556-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...