5,7-Difluorchinolin 95%

Lieferant: Apollo Scientific

PC906597-1G
APOSPC906597-1GEA 108 EUR
APOSPC906597-1G
5,7-Difluorchinolin 95%
5,7-Difluoroquinoline
Formel: C₉H₅F₂N
Molecular Weight: 165.1400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00474638
CAS-Nummer: 34522-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR