3-Hydroxy-4-(trifluoromethyl)benzonitrile 95%
Lieferant: Apollo Scientific
PC906689-1G
APOSPC906689-1GEA
82.8
EUR
APOSPC906689-1G
3-Hydroxy-4-(trifluoromethyl)benzonitrile 95%
3-Hydroxy-4-(trifluoromethyl)benzonitrile
Formel:
C₈H₄F₃NO Molecular Weight: 187,119 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16999170 CAS-Nummer: 731002-50-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...