2-Fluoro-1,3-dimethyl-4-nitrobenzene 95%
Lieferant: Apollo Scientific
PC906703-5G
APOSPC906703-5GEA
128
EUR
APOSPC906703-5G
2-Fluoro-1,3-dimethyl-4-nitrobenzene 95%
2-Fluoro-1,3-dimethyl-4-nitrobenzene
Formel:
C₈H₈FNO₂ Molecular Weight: 169,153 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22683872 CAS-Nummer: 1736-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...