7-Trifluoromethyl-imidazo[1,2-a]pyridine 95%

Lieferant: Apollo Scientific

PC906729-1G PC906729-250MG
APOSPC906729-1GEA 632 EUR
APOSPC906729-1G APOSPC906729-250MG
7-Trifluoromethyl-imidazo[1,2-a]pyridine 95%
7-Trifluoromethyl-imidazo[1,2-a]pyridine
Formel: C₈H₅F₃N₂
Molecular Weight: 186.134 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035989
CAS-Nummer: 944580-91-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR