2,4-Dichloro-6-fluorobenzonitrile 95%

Lieferant: Apollo Scientific

PC906758-1G
APOSPC906758-1GEA 246 EUR
APOSPC906758-1G
2,4-Dichloro-6-fluorobenzonitrile 95%
2,4-Dichloro-6-fluorobenzonitrile
Formel: C₇H₂Cl₂FN
Molecular Weight: 190,002 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19686188
CAS-Nummer: 1349718-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR