2,6-Difluoro-4-propylphenylboronic acid 95%

Lieferant: Apollo Scientific

PC906796-1G
APOSPC906796-1GEA 632 EUR
APOSPC906796-1G
2,6-Difluoro-4-propylphenylboronic acid 95%
2,6-Difluoro-4-propylphenylboronic acid
Formel: C₉H₁₁BF₂O₂
Molecular Weight: 199,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21337851
CAS-Nummer: 1010110-74-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR