2,4-Dichlor-6-fluorbenzaldehyd 95%

Lieferant: Apollo Scientific

PC906832-1G
APOSPC906832-1GEA 46.4 EUR
APOSPC906832-1G
2,4-Dichlor-6-fluorbenzaldehyd 95%
2,4-Dichloro-6-fluorobenzaldehyde
Formel: C₇H₃Cl₂FO
Molecular Weight: 193,0000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12404935
CAS-Nummer: 681435-09-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR