3-Amino-2,5,6-trifluorobenzonitrile 95%

Lieferant: Apollo Scientific

PC906853-1G
APOSPC906853-1GEA 647 EUR
APOSPC906853-1G
3-Amino-2,5,6-trifluorobenzonitrile 95%
3-Amino-2,5,6-trifluorobenzonitrile
Formel: C₇H₃F₃N₂
Molecular Weight: 172,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20717606
CAS-Nummer: 159847-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR