(3-Fluoroazetidin-3-yl)methanol

Lieferant: Apollo Scientific
Achtung

PC906910-1G
APOSPC906910-1GEA 877 EUR
APOSPC906910-1G
(3-Fluoroazetidin-3-yl)methanol
(3-Fluoroazetidin-3-yl)methanol
Formel: C₄H₈FNO
Molecular Weight: 105,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18633069
CAS-Nummer: 1268520-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR