1-Fluoro-2,3-dimethoxybenzene 95%
Lieferant: Apollo Scientific
PC906944-1G
APOSPC906944-1GEA
39.3
EUR
APOSPC906944-1G
1-Fluoro-2,3-dimethoxybenzene 95%
1-Fluoro-2,3-dimethoxybenzene
Formel:
C₈H₉FO₂ Molecular Weight: 156,154 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03411451 CAS-Nummer: 394-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...