2-Bromo-1,5-difluoro-3-nitrobenzene 95%

Lieferant: Apollo Scientific

PC906985-5G
APOSPC906985-5GEA 173 EUR
APOSPC906985-5G
2-Bromo-1,5-difluoro-3-nitrobenzene 95%
2-Bromo-1,5-difluoro-3-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD04112512
CAS-Nummer: 877161-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR