4-Fluoropiperidine-4-carbonitrile hydrochloride ≥98%

Lieferant: Apollo Scientific

PC906999-100MG
APOSPC906999-100MGEA 580 EUR
APOSPC906999-100MG
4-Fluoropiperidine-4-carbonitrile hydrochloride ≥98%
4-Fluoropiperidine-4-carbonitrile hydrochloride
Formel: C₆H₉FN₂·ClH
Molecular Weight: 164,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19689455
CAS-Nummer: 1374653-45-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR