4-(Chloromethyl)-2-[4-(trifluoromethyl)phenyl]thiazole 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
PC9070-250MG
APOSPC9070-250MGEA
0
EUR
APOSPC9070-250MG
4-(Chloromethyl)-2-[4-(trifluoromethyl)phenyl]thiazole 95+%
4-(Chloromethyl)-2-[4-(trifluoromethyl)phenyl]thiazole
Formel:
C₁₁H₇ClF₃NS Schmelzpunkt: 80…82 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00277208 CAS-Nummer: 135873-35-7 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...