3-Fluoroazetidine-3-carboxylic acid

Lieferant: Apollo Scientific

PC907021-1G
APOSPC907021-1GEA 1490 EUR
APOSPC907021-1G
3-Fluoroazetidine-3-carboxylic acid
3-Fluoroazetidine-3-carboxylic acid
Formel: C₄H₆FNO₂
Molecular Weight: 119,094 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22566212
CAS-Nummer: 1363380-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR