6,7-Difluorochroman-4-one 95%

Lieferant: Apollo Scientific

PC907037-1G
APOSPC907037-1GEA 184 EUR
APOSPC907037-1G
6,7-Difluorochroman-4-one 95%
6,7-Difluorochroman-4-one
Formel: C₉H₆F₂O₂
Molecular Weight: 184,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11518477
CAS-Nummer: 1092349-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR