5-Fluoro-2-methylbenzotrifluoride 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-1-methyl-2-(trifluoromethyl)benzene
PC907074-1G
PC907074-25G
PC907074-5G
APOSPC907074-1GEA
27.7
EUR
APOSPC907074-1G
APOSPC907074-25G
APOSPC907074-5G
5-Fluoro-2-methylbenzotrifluoride 99%
5-Fluoro-2-methylbenzotrifluoride
Formel:
C₈H₆F₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11226544 CAS-Nummer: 141872-92-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...