5-Chloro-3-fluorobenzene-1,2-diol 95%

Lieferant: Apollo Scientific

PC907077-1G
APOSPC907077-1GEA 647 EUR
APOSPC907077-1G
5-Chloro-3-fluorobenzene-1,2-diol 95%
5-Chloro-3-fluorobenzene-1,2-diol
Formel: C₆H₄ClFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18382509
CAS-Nummer: 645405-05-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR