2-Fluoro-4-nitrophenylacetonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(2-Fluoro-4-nitrophenyl)acetonitrile
PC907083-1G
APOSPC907083-1GEA
91.5
EUR
APOSPC907083-1G
2-Fluoro-4-nitrophenylacetonitrile
2-Fluoro-4-nitrophenylacetonitrile
Formel:
C₈H₅FN₂O₂ Molecular Weight: 180,136 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD13185637 CAS-Nummer: 853910-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...