2-Fluoro-5-methoxy-4-methylbenzoic acid 95%

Lieferant: Apollo Scientific

PC907086-5G
APOSPC907086-5GEA 2840 EUR
APOSPC907086-5G
2-Fluoro-5-methoxy-4-methylbenzoic acid 95%
2-Fluoro-5-methoxy-4-methylbenzoic acid
Formel: C₉H₉FO₃
Molecular Weight: 184,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440286
CAS-Nummer: 870221-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR