2-Fluoro-5-hydroxy-4-methylbenzoic acid 95%
Lieferant: Apollo Scientific
PC907093-5G
APOSPC907093-5GEA
339
EUR
APOSPC907093-5G
2-Fluoro-5-hydroxy-4-methylbenzoic acid 95%
2-Fluoro-5-hydroxy-4-methylbenzoic acid
Formel:
C₈H₇FO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD19440271 CAS-Nummer: 870221-14-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...