4'-Fluor-2-methylpropiophenon

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Fluoroisobutyrophenone, 1-(4-Fluorophenyl)-2-methylpropan-1-one

PC9071-1G PC9071-25G PC9071-5G
APOSPC9071-1GEA 69.6 EUR
APOSPC9071-1G APOSPC9071-25G APOSPC9071-5G
4'-Fluor-2-methylpropiophenon
4'-Fluor-2-methylpropiophenon
Formel: C₁₀H₁₁FO
Molecular Weight: 166.19 g/mol
Siedepunkt: 95…97 °C/0.7mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04038305
CAS-Nummer: 26393-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR